Would you like to log in again?
B-Phycoerythrin (BPE), Columbia Biosciences
Supplier: Columbia Biosciences
D4-000
10065-812EA 45.42 USD InStock
10065-812
B-Phycoerythrin (BPE), Columbia Biosciences
Protein Linking and Labeling Systems
Because of its high quantum yield, B-PE is considered the world’s brightest fluorophore
It is compatible with commonly available lasers and gives exceptional results in flow cytometry, Luminex® and immunofluorescent staining. B-PE is also less “sticky” than common synthetic fluorophores and therefore gives less background interference.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty