Would you like to log in again?
ACTERO™ Listeria Enrichment Media, FoodChek Systems
Supplier: FOODCHEK SYSTEMS INC.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FCM-101 FCM-011 FCM-037 FCM-055 FCM-083 FCM-039 FCM-104 FCM-056 FCM-162 FCM-084 FCM-084 FCM-056 FCM-213 FCM-102 FCM-011 FCM-137 FCM-105 FCM-057 FCM-057 FCM-022 FCM-037 FCM-161 FCM-163 FCM-038 FCM-038 FCM-052 FCM-054 FCM-053 FCM-082 FCM-100 FCM-162 FCM-100 FCM-103 FCM-054 FCM-211 FCM-082 FCM-210 FCM-213 FCM-139 FCM-138 FCM-102 FCM-211 FCM-137 FCM-022 FCM-053 FCM-161 FCM-052 FCM-103 FCM-039 FCM-023 FCM-104 FCM-101 FCM-210 FCM-138 FCM-083 FCM-139 FCM-055 FCM-163
10065-990EA 339.84 USD InStock
10065-990 CA89522-694 CA89522-692 10065-992 CA89522-690 10146-990 10146-670 10159-090 10066-070 10066-072 CA10158-484 10066-068 CA97068-830 CA10066-830 CA10066-676 CA10066-832 CA10066-824 10146-668 CA10066-826 10065-982 CA10066-828 10065-986 10065-984 CA10158-514 10065-988 CA10158-512 CA10158-510 10066-052 CA10158-476 97068-830 CA10158-474 CA10158-472 CA10066-664 CA10066-666 CA10066-668 CA10066-822 10146-654 10146-698 10146-694 10146-696 10066-090 10066-050 103257-596 CA10158-860 10066-048 CA10066-850 CA10066-852 CA10066-854 CA10066-846 CA10066-848 89522-694 89522-690 10159-084 89522-692 10159-086 10066-080 CA10158-856 CA10158-854
ACTERO™ Listeria Enrichment Media, FoodChek Systems
Media for Microbiology Prepared Media for Microbiology
Food and environmental samples require enrichment before pathogen testing enabling targetl pathogen present to grow and be detected in test samples
Using Actero™ Elite Enrichment Media significantly reduces all sample enrichment phase timelines regardless the type of "food pathogen testing system," such as lateral flow, ELISA and PCR platforms.
Actero™ Universal Enrichment Media are established formulations that are used in standard protocols employed in testing labs today.
The patented Actero™ Elite Enrichment Media are enhanced formulations that reduce the enrichment time required for accurate pathogen testing. Actero™ Listeria Enrichment Media recieved the AOAC-PTM (111201) accreditation to be used as a single-step recovery and enrichment of Listeria spp., including Listeria monocytogenes from selected food and environmental samples.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty