Would you like to log in again?
Silylation Grade Solvents
Supplier: Thermo Scientific
TS-20062 TS-27530 TS-20672
10066-272EA 243.56 USD InStock
10066-272 10066-268 10066-292
Silylation Grade Solvents
Chromatography Sorbents GC Sorbents
Use specially manufactured and packaged Silylation Grade Solvents to meet the exacting needs of silylation and other sensitive derivatization reactions.
  • Purified, dried, and packaged under nitrogen in convenient Hypo-Vial™ sample storage vials,
  • Supplied with elastomer septa to enable immediate access to the sample without exposure to moisture and oxygen
Recommended use: Avoid water or alcohol, as TMS reagents react with active hydrogen. Avoid enolizable ketones. Use dimethylformamide for steroids and other large molecules. Use dimethylsulfoxide to prepare TMS derivatives of tertiary alcohols and some compounds with low solubility in other silylation solvents.
Use polar solvents (acetonitrile, dimethylformamide, dimethylsulfoxide, pyridine, tetrahyrdrofuran) to facilitate reactions; nonpolar organic solvents may be used but will not accelerate the rate of reaction. Pyridine is an excellent solvent and reaction medium for MS reactions and is an HCI acceptor in reactions involving organochlorosilanes.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty