Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BSTFA Silylation Reagents

Supplier: Thermo Scientific
TS-38829 TS-38828 TS-38830
10066-310EA 581.92 USD InStock
10066-310 10066-312 10066-308
BSTFA Silylation Reagents
Chromatography Sorbents GC Sorbents
Improve GC analysis with Thermo Scientific™ BSTFA Silylation Reagent. The highly volatile byproducts mono(trimethylsilyl)trifluoroacetamide (BSTFA) and trifluoroacetamide elute with the solvent front to ensure excellent chromatographic separations.

  • Increased volatility
  • Store at 2 to 8 °C
  • Standard concentration: 98 to 100%
  • Standard components: Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate

This silylation reagent reacts with a wide range of polar compounds and is particularly well suited for the GC analyses of some lower boiling TMS-amino acids and TMS Krebs cycle acids.

BSTFA is a powerful thrimethylsilyl donor, with donor strength that is comparable to its unfluorinated analog BSA [N,O-Bis(trimethylsilyl)acetamide]. BSTFA reacts to replace labile hydrogens on a wide range of polar compounds with a Si(CH₃)₃ group.

Increased volatility of reaction byproducts mono(trimethylsilyl)trifluoroacetamide and trifluoroacetamide over corresponding nonfluorinated compounds from BSA. Increased volatility makes it possible to derivatize smaller molecules with which the TMS derivatives elute with the byproducts from BSA.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty