Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tri-Sil TP (TMSI:Pyridine) Reagents

Supplier: Thermo Scientific
TS-49230 TS-49231
10066-362EA 423.04 USD InStock
10066-362 10066-364
Tri-Sil TP (TMSI:Pyridine) Reagents
Chromatography Sorbents GC Sorbents
Derivatize hydroxyls compounds, particularly carbohydrates with Tri-Sil TP (TMSI:Pyridine) reagent.

  • Does not react with amines
  • May be used with water as long as there is enough reagent present to react with both the water and the sample
  • For use with GC
  • Standard concentration: 65 to 80%, 20 to 25%
  • Standard components are Pyridine, 1-(Trimethylsilyl)-1 H-imidazole
  • Storage at 2 to 8 °C

Comprising TMSI in pyridine, this reagent silylates alcohols and phenols, organic acids, hydroxylamines, amino acids, carbohydrates, flavonoids, glycols and polyglycols, nucleotides, steroids, hydroxyl acids, barbituates, narcotics, indoles, and vitamins.

Caution: For research use only. Not for use in diagnostic procedures.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty