Would you like to log in again?
PFFA 10 X 1ML AMP
Supplier: Thermo Scientific
PFAA^ PFFA 10 X 1ml ampule, high-purity, ideal for preparing fluoracyl derivatives, used to prepare fluoracyl derivatives for GC/MS, they produce stable volatile derivatives for FID and ECD techniques
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PFFA 10 X 1ML AMP 10066-400 TS-65193 Each (1ml) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty