Would you like to log in again?
Silylation Reagent, HMDS (Hexamethyldisilazane)
Supplier: Thermo Scientific
TS-84770
10066-406EA 439.29 USD InStock
10066-406
Silylation Reagent, HMDS (Hexamethyldisilazane)
Chromatography Sorbents GC Sorbents
HMDS (Hexamethyldisilazane) greatly extends the practical range of GC, improving chromatographic results in the silylation of sugars and related substances.
  • Suitable for deactivating and coating chromatographic supports
  • Monofunctional, making polymerization not possible and eliminating surface moisture
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty