Would you like to log in again?
Tweezers, Acetal Soft Replaceable Tip, Excelta Corp®
Supplier: Excelta
159D-RTW 159B-RTW 159C-RTW 159D-RTW 159C-RTW 759D-RTW 759-RTW
76238-652EA 91.9 USD InStock
76238-652 94024-110 76241-090 94024-114 76471-584 10066-506 76471-378
Tweezers, Acetal Soft Replaceable Tip, Excelta Corp®
Forceps
The acetal tips are specifically used for handling items that cannot be scratched, like contact lenses.
  • Will not scratch or mar parts
  • Available in several tip widths
  • Cleanroom safe, will not particulate
  • Resistant to most chemicals and solvents
These straight replaceable tip tweezers are 5" long and are used primarily to prevent marring or scratching of delicate parts. Acetal has outstanding resistance to chemicals and solvents. They are electrically insulative and resistant to temperatures up to 185 °F. The bodies are made of anti-magnetic Neverust® stainless steel (AISI 316L). Excellent for cleanroom applications and for avoiding the marring or scratching of items being handled. The replacement tips include 4 screws and 2 tips.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty