Would you like to log in again?
LIM Broth, for Group B Strep
Supplier: Hardy Diagnostics
L57
10066-596PK 114.98 USD InStock
10066-596
LIM Broth, for Group B Strep
Media for Microbiology Prepared Media for Microbiology
LIM Broth is recommended for the selective enrichment of group B streptococci (Streptococcus agalactiae) from anovaginal specimens from pregnant women.
  • Flat bottom contributes to stability when set on end
  • Leak-proof caps provide security during handling and transportation
  • Robust construction tubes have excellent impact and are shatter resistant
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty