Would you like to log in again?
Sticky Mat Refills, Ergomat
Supplier: Ergomat
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SM1504-BLU SM1504-WHITE
10067-688EA 335.75 USD InStock
10067-688 10067-686
Sticky Mat Refills, Ergomat
Floor Mats Tacky Mats
Ergomat Sticky Mats provide a fast, convenient, reliable way to clean shoe soles or other items before entering a clean and sanitary area
  • 30 layers per mat
  • Easily adhered to a frame with double-faced adhesive tape
  • Mats utilize a water-based acrylic adhesive, which has a pH of 7–8
  • A clear release sheet protects the top sheet of the sticky mat until it is ready to use
  • Corner tabs are numbered to show the number of remaining sheets
These significantly reduce traffic-borne contaminants and have proven themselves effective in a wide variety of industries such as microelectronics, aerospace, pharmaceuticals, and food processing. The disposable sticky mat consists of a stack of tear-off polyethylene films full of adhesive sheets that effectively capture dirt and dust.
Certifications: Meets DIN/ASTM requirements.
Ordering information: Frame must be ordered separately.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty