Would you like to log in again?
Optimus™ Combination Emergency Shower and Eye/Face Wash System, Speakman®
Supplier: Speakman
Optimus™
SE-1200 SE-1250 SE-1255
10068-222EA 2819.32 USD InStock
10068-222 10068-224 10068-220
Optimus™ Combination Emergency Shower and Eye/Face Wash System, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Combination emergency shower and eye/face wash system equipped with a dual aerated emergency eye wash to deliver soothing relief.
  • Combined with emergency shower with 8" Ø shower head
  • Two individual face pads producing 66 individual, full-coverage sprays
  • Three year limited warranty
Features a highly visible yellow ABS plastic or corrosion resistant stainless steel bowl attached to a powder-coated steel emergency shower.
Certifications: ANSI/ISEA Z358.1 certified, OSHA compliant. Pipe fittings can be customized to meet ADA height specifications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty