Would you like to log in again?
Human Recombinant Myostatin Propeptide (from E. coli)
Supplier: ProSci
40-210
10072-762EA 302.43 USD InStock
10072-762
Human Recombinant Myostatin Propeptide (from E. coli)
Proteins and Peptides
Mature Myostatin is obtained by proteolytic processing of a biologically-inactive precursor protein, which contains an N-terminal propeptide of 243 amino acid residues. Myostatin Propeptide exhibits high binding affinity for myostatin and has been shown to be a potent inhibitor of Myostatin. Over-expression of myostatin propeptide in mice resulted in large increases (up to 200%) in skeletal muscle mass, similar to those observed in Myostatin knockout mice. Recombinant Human Myostatin Propeptide is a 27.8 kDa protein consisting of 244 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty