Would you like to log in again?
Human Recombinant IGFBP6
Supplier: ProSci
40-320
10072-934EA 302.43 USD InStock
10072-934
Human Recombinant IGFBP6
Proteins and Peptides
IGF-BPs controls the distribution, function and activity of IGFs in various cell tissues and body fluids. IGF-BP6 is produced by bone cells and is the major IGF-BP present in cerebrospinal fluid, and specifically inhibits IGF-II actions. IGF-BP6 has been shown to inhibit IGF-II-dependent cancers such as neuroblastoma, colon cancer and rhabdomyosarcoma. Recombinant human IGF-BP6 has a calculated mass of 22.6 kDa and consists of 213 amino acid residues including the IGF-BP domain and thyroglobulin type-I domain. IGF-BP6 migrates at an apparent molecular weight of approximately 23.0-30.0 kDa by SDS-PAGE analysis under non-reducing conditions.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty