Would you like to log in again?
Biocide Antimicrobial Dry Vac, Atrix
Supplier: Atrix
ATIBCV
100720-962EA 1088.68 USD InStock
100720-962
Biocide Antimicrobial Dry Vac, Atrix
Vacuum Cleaners
The Antimicrobial Dry Vacuum Cleaner has all plastic components and attachments containing Microban to prevent proliferation of bacteria and viruses
Two HEPA filters are used within the Antimicrobial Dry Vacuum Cleaner. The primary filter catches the first of the incoming particles and a secondary filter prevents escape of dust from the carbon brushes. For added convenience an indicator light alerts the operator when to change the bag
It uses a powerful 1,000 Watt super high speed motor for deep cleaning. While powerful, the Antimicrobial Dry Vacuum Cleaner is also extra quite for sound sensitive environments such as human and animal hospitals, nursing facilites, and in-home care. Two HEPA filters are used within the Antimicrobial Dry Vacuum Cleaner. The primary filter catches the first of the incoming particles and a secondary filter prevents escape of dust from the carbon brushes. For added convenience an indicator light alerts the operator when to change the bag.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty