Would you like to log in again?
Mycoplasma crocodyli Recombinant Mycoplasma Arginine Deiminase (from E. coli)
Supplier: ProSci
40-582
10073-066EA 302.43 USD InStock
10073-066
Mycoplasma crocodyli Recombinant Mycoplasma Arginine Deiminase (from E. coli)
Enzymes
Arginine Deiminase (ADI) is a microbial enzyme from Mycoplasma produced in E.coli
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
It has high affinity to L-arginine and hydrolyzes L-arginine to citrulline and ammonia. Low concentrations of ADI have been shown to inhibit proliferation in certain cultured cells by arresting the cell cycle in G (1) and/or S phase. Higher concentrations of ADI lead to subsequent apoptosis. Recombinant Mycoplasma Arginine Deiminase is a 46.4 kDa protein consisting of 409 amino acids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty