Would you like to log in again?
Rat Recombinant Carboxypeptidase-B (from E. coli)
Supplier: ProSci
40-583
10073-068EA 302.43 USD InStock
10073-068
Rat Recombinant Carboxypeptidase-B (from E. coli)
Enzymes
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Most proteases act in a specific manner, hydrolyzing bonds at or adjacent to specific residues or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Carboxypeptidase-B sequentially cleaves C terminal K and R residues. Recombinant rat Carboxypeptidase-B is a 35.1 kDa protein consisting of 307 amino acids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty