Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TMEM27 Rabbit Polyclonal Antibody

Supplier: Proteintech
19207-1-AP-150UL
10081-170EA 453 USD InStock
10081-170
Anti-TMEM27 Rabbit Polyclonal Antibody
Antibodies
TMEM27(Transmembrane protein 27) is also named as collectrin and Belongs to the TMEM27 family .It is a stimulator of beta cell replicationa and a membrane protein cleaved and shed by pancreatic beta cells that has been proposed as a beta cell mass biomarker.Western blot analysis of mouse collecting duct cell lines detected Tmem27 at an apparent molecular mass of 32 kD(11278314).

Western Blot: Mouse pancreas Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: TMEM27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty