Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ENTPD8 Rabbit Polyclonal Antibody

Supplier: Proteintech
21010-1-AP-150UL
10081-296EA 453 USD InStock
10081-296
Anti-ENTPD8 Rabbit Polyclonal Antibody
Antibodies
ENTPD8(Ectonucleoside triphosphate diphosphohydrolase 8) is also named as E-NTPDase 8, NTPDase8 and belongs to the GDA1/CD39 NTPase family. NTPDase8 is the canalicular ecto-ATPase/ATPDase and is responsible for the main hepatic NTP-Dase activity. The estimated molecular mass of expressed human E-NTPDase 8 is 70-85 kDa which is N-glycosylated and after deglycosylation,it has a nearly identical molecular mass, 54 kDa. It has 2 isoforms produced by alternative splicing with the molecular weight of 54 kDa and 49 kDa. This antibody is specific to ENTPD8.

Western Blot: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: ENTPD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty