Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VPS45 Rabbit Polyclonal Antibody

Supplier: Proteintech
12006-1-AP-150UL
10081-342EA 453 USD InStock
10081-342
Anti-VPS45 Rabbit Polyclonal Antibody
Antibodies
The Sec1/Munc18 (SM) family of proteins regulate intracellular trafficking through interactions with individual SNARE (soluble N-ethylmeleimide-sensitive factor attachment protein receptor) proteins and assembled SNARE complexes. The SM protein VPS45 (Vacuolar protein sorting-associated protein 45) has a calculated molecular mass of 65 kDa and shares 38% identity with yeast Vps45 which is required for the formation of yeast endosomal SNARE complexes and is thus essential for traffic through the endosomal system. VPS45 may play a role in vesicle-mediated protein trafficking from the Golgi stack through the trans-Golgi network.

IHC: Human gliomas Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: VPS45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty