Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACPP Mouse Monoclonal Antibody [clone: 1G2C9]

Supplier: Proteintech
60176-1-IG-150UL
10081-892EA 453 USD InStock
10081-892
Anti-ACPP Mouse Monoclonal Antibody [clone: 1G2C9]
Antibodies
ACPP is also named as PAP(prostatic acid phosphatase),5'-nucleotidase,thiamine monophosphatas and Belongs to the histidine acid phosphatase family.This enzyme is a homodimeric glycoprotein with a subunit molecular mass of 50 kDa and is an androgen-stimulated secretory product of the human prostate gland.It is also a non-specific phosphatase and has been implicated in the progression of prostate cancer, which inactivates LPA in human seminal plasma. This protein has three glycosylation sites.

Western Blot: Human Testis, 1:200-1:1000; IHC: Human prostate hyperplasia Tissue, 1:20-1:200

Type: Primary
Antigen: ACPP
Clonality: Monoclonal
Clone: 1G2C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty