Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CTSD Rabbit Polyclonal Antibody

Supplier: Proteintech
55021-1-AP-150UL
10082-000EA 453 USD InStock
10082-000
Anti-CTSD Rabbit Polyclonal Antibody
Antibodies
CTSD(Cathepsin D) is also named as CPSD,belongs to the peptidase A1 family.It is ubiquitously expressed and is involved in proteolytic degradation, cell invasion, and apoptosis. The prepro- CTSD isoform has a molecular mass of 52 kDa, the pro-CTSD 48 kDa and the mature CTSD 32 kDa. It is a lysosomal acid protease found in neutrophils and monocytes and involved in the pathogenesis of several diseases such as breast cancer and possibly Alzheimer disease.Defects in CTSD are the cause of neuronal ceroid lipofuscinosis type 10 (CLN10).

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: CTSD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty