Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACADVL Rabbit Polyclonal Antibody

Supplier: Proteintech
14527-1-AP-150UL
10082-400EA 453 USD InStock
10082-400
Anti-ACADVL Rabbit Polyclonal Antibody
Antibodies
VVery long-chain acyl-CoA dehydrogenase (VLCAD) is one of four flavoproteins which catalyze the initial step of the mitochondrial b-oxidation spiral. It belongs to the acyl-CoA dehydrogenase family and is a homodimer of a 71-kDa polypeptide which contains 2 mol of FAD/mol of enzyme. The molecular mass of the nondenatured trypsinized VLCAD is 98 kDa, by gel filtration chromatography, indicating that it is a homodimer of the 48 kDa(tryptic digest) polypeptide. Defects in ACADVL are the cause of acyl-CoA dehydrogenase very long chain deficiency (ACADVLD). It has 2 isoforms(70 kDa and 68 kDa ) produced by alternative splicing and a transit peptide. This antibody is speicifc to ACADVL.

Western Blot: Mouse Liver Tissue, 1:200-1:2000; IHC: Human Liver cancer, 1:20-1:200

Type: Primary
Antigen: ACADVL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty