Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACP5 Rabbit Polyclonal Antibody

Supplier: Proteintech
11594-1-AP-150UL
10082-450EA 453 USD InStock
10082-450
Anti-ACP5 Rabbit Polyclonal Antibody
Antibodies
ACP5(acid phosphatase 5, tartrate resistant) is also named as TRAP, TrATPase and belongs to the metallophosphoesterase superfamily. It is an iron-containing glycoprotein with a molecular mass of approximately 34 kD. ACP5 is a histochemical marker of the osteoclast. It is also characteristic of monohistiocytes, particularly alveolar macrophages, and is associated with diverse pathological conditions, including hairy cell leukemia and AIDS encephalopathy..

IHC: Human breast cancer Tissue, 1:10-1:100

Type: Primary
Antigen: ACP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty