Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ASPA Rabbit Polyclonal Antibody

Supplier: Proteintech
13244-1-AP-150UL
10083-204EA 453 USD InStock
10083-204
Anti-ASPA Rabbit Polyclonal Antibody
Antibodies
ASPA(Aspartoacylase) also called aminoacylase-2, is an enzyme that hydrolyzes N-acetyl-L-aspartic acid (NAA) to aspartate and acetate.Human ASPA encodes a deduced 313-amino acid protein with a molecular mass of 36 KDa. It shares 92% sequence identity with the bovine protein and contains 1 potential N-glycosylation site and 5 phosphorylation sites.Defects in ASPA are the cause of Canavan disease (CAND).This protein can exsit as a homodimer(18293939).

Western Blot: Mouse Kidney Tissue, 1:200-1:2000; IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: ASPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty