Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP8 Rabbit Polyclonal Antibody

Supplier: Proteintech
13423-1-AP-150UL
10084-096EA 453 USD InStock
10084-096
Anti-CASP8 Rabbit Polyclonal Antibody
Antibodies
CASP8, also named as MCH5, MACH, FLICE, and CAP4, belongs to the peptidase C14A family. It may participate in the GZMB apoptotic pathways and functions as an upstream regulator in a-bisabolol-induced apoptosis. CASP8 catalyzes an essential intermediate step in the ubiquitination and proteasome-mediated degradation of IRF3. It may control diabetic embryopathy-associated apoptosis via regulation of the Bid-stimulated mitochondrion/caspase-9 pathway. CASP8 is expressed as nine isoforms by alternative splicing with the molecular mass from 26 kDa to 62 kDa.

Western Blot: HepG2 Cells, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200; IP: HeLa Cells, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CASP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty