Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDK9 Rabbit Polyclonal Antibody

Supplier: Proteintech
11705-1-AP-150UL
10084-534EA 453 USD InStock
10084-534
Anti-CDK9 Rabbit Polyclonal Antibody
Antibodies
CDK9(Cyclin-dependent kinase 9) is involved in the regulation of transcription.It has two isforms.CDK9-55 had an apparent molecular mass of 55 kD. The ratio of CDK9-55 to the shorter CDK9 isoform, CDK9-42, differed between HeLa cell nuclear extracts and mouse fibroblasts, but CDK9-42 was the predominant isoform in both.CDK9 is located primarily in the nucleus.

Western Blot:A431 Cells, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: CDK9
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty