Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Centrin 2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15877-1-AP-150UL
10084-582EA 453 USD InStock
10084-582
Anti-Centrin 2 Rabbit Polyclonal Antibody
Antibodies
Centrin 2 is also named as CALT, or CEN2. It plays a fundamental role in microtubule-organizing center structure and function. Oxidizing radiation induces polymerization of centrin 2 into dimers, tetramers, hexamers,and even higher molecular mass species, as a function of the radiation dose.It exsits as a 21-kD polypeptide specifically localized to the centrosome of interphase and mitotic cells in both HeLa and BHK cells .

IHC: Human Brain Tissue, 1:20-1:200; Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: Centrin 2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty