Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYP17A1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14447-1-AP-150UL
10085-498EA 453 USD InStock
10085-498
Anti-CYP17A1 Rabbit Polyclonal Antibody
Antibodies
The human cytochrome P450c17 enzyme is encoded by the CYP17A1 gene. It is the qualitative regulator of steroidogenesis in the human adrenals.In its absence, aldosterone is produced in the human adrenal zona glomerulosa .The CYP17A1 gene is expressed in the adrenal cortex and the gonads but not in the placenta . Although CYP17A1 has been shown to be a 50-kDa protein, the presence of a lower molecular mass(30 kDa) immunoreactive form in rat Ley- dig cells was previously shown and it is either a degradation product or a truncated form of the 50 kDa enzyme. Defects in CYP17A1 are the cause of adrenal hyperplasia type 5 (AH5).

Western Blot: HeLa Cells, 1:200-1:1000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: CYP17A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty