Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FA2H Rabbit Polyclonal Antibody

Supplier: Proteintech
15452-1-AP-150UL
10086-754EA 453 USD InStock
10086-754
Anti-FA2H Rabbit Polyclonal Antibody
Antibodies
FA2H(Fatty acid 2-hydroxylase) required for alpha-hydroxylation of free fatty acids and the formation of alpha-hydroxylated sphingolipids. The deduced 372-amino acid protein has a calculated molecular mass of 42.8 kD .Both mouse and human FA2H have a C-terminal endoplasmic reticulum (ER) retention motif.

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: FA2H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty