Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HEXB Rabbit Polyclonal Antibody

Supplier: Proteintech
16229-1-AP-150UL
10087-288EA 453 USD InStock
10087-288
Anti-HEXB Rabbit Polyclonal Antibody
Antibodies
The HEXB(Beta-hexosaminidase subunit beta) gene encodes the beta subunit of the enzyme hexosaminidase, which is involved in the breakdown of gangliosides. It is responsible for the degradation of GM2 gangliosides, and has a variety of other molecules containing terminal N-acetyl hexosamines, in the brain and other tissues.It has a calculated molecular mass of about 63 KDa.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: HEXB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty