Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LIAS Rabbit Polyclonal Antibody

Supplier: Proteintech
11577-1-AP-150UL
10088-228EA 453 USD InStock
10088-228
Anti-LIAS Rabbit Polyclonal Antibody
Antibodies
LIAS(lipoyl synthase, mitochondrial) is also named as LAS and belongs to the radical SAM superfamily and lipoyl synthase family. It produces alpha-lipoic acid, an antioxidant and an essential cofactor in alpha-ketoacid dehydrogenase complexes, which participate in glucose oxidation and ATP generation. The deduced 373-amino acid protein has a calculated molecular mass of about 42 kD. The N-terminal 26 amino acids encode a potential mitochondrial targeting presequence that, upon removal, would result in a deduced mature protein of 347 amino acids with a molecular mass of about 39 kD. Defects in LIAS are a cause of pyruvate dehydrogenase lipoic acid synthetase deficiency (PDHLD).

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: LIAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty