Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITPKB Rabbit Polyclonal Antibody

Supplier: Proteintech
12816-1-AP-150UL
10089-048EA 453 USD InStock
10089-048
Anti-ITPKB Rabbit Polyclonal Antibody
Antibodies
Inositol 1,4,5-trisphosphate 3-kinase isoform B (IITPKB) is also named as IP3KB and possesses important roles in the development of immune cells. IP3KB can be targeted to multiple cellular compartments, among them nuclear localization and binding in close proximity to the plasma membrane. The B isoform is the only IP3K that is almost ubiquitously expressed in mammalian cells.Three isoforms of IP3KB occur in mammals, which are distinguished by different molecular masses, sensitivity to Ca2+/CaM(calmodulin), intracellular distribution and tissue expression .

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human cervical cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ITPKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty