Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LMBR1L Rabbit Polyclonal Antibody

Supplier: Proteintech
12154-1-AP-150UL
10089-728EA 453 USD InStock
10089-728
Anti-LMBR1L Rabbit Polyclonal Antibody
Antibodies
LMBR1L, also known as LIMR (lipocalin-1-interacting membrane receptor), interacts with lipocalin-1 (Lcn-1), a lipocalin member produced by a number of secretoric glands and tissues and known to bind a variety of lipophilic compounds. LMBR1L is a 489-amino acid protein with a predicted molecular mass of 55 kD and 9 putative transmembrane helices. LMBR1L has been shown to mediate cellular uptake of LMBR1L, thus it is characterized as a endocytic receptor.

Western Blot: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: LMBR1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty