Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MDM4 Rabbit Polyclonal Antibody

Supplier: Proteintech
17914-1-AP-150UL
10090-072EA 453 USD InStock
10090-072
Anti-MDM4 Rabbit Polyclonal Antibody
Antibodies
MDM4 is also named as MDMX. It inhibits the activity of the tumor suppressor p53 by primarily cooperating with the p53 feedback regulator MDM2. MDM4 resides mostly in the cytoplasm, but can be recruited to the nucleus by MDM2.. The predicted mass of the protein is 54 kD, while the observed mass is 80 kD, a difference which stated is probably due to phosphorylation or other posttranslational modification. It can be ubiquitinated and degraded by MDM2. It has 5 isoforms produced by alternative splicing.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: MDM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty