Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MED20 Rabbit Polyclonal Antibody

Supplier: Proteintech
17598-1-AP-150UL
10090-092EA 453 USD InStock
10090-092
Anti-MED20 Rabbit Polyclonal Antibody
Antibodies
MED20(Mediator of RNA polymerase II transcription subunit 20) is also named as TRFP and belongs to the Mediator complex subunit 20 family. The deduced 209-amino acid protein shares 44% amino identity with Drosophila Trfp. TRFP had an apparent molecular mass of 28 kD by SDS-PAGE. It is identified TRFP as a component of an RNA polymerase II-SRB complex. The TRFP-containing complex could substitute for core RNA polymerase II in driving basal transcription from a minimal promoter.

Western Blot:K-562 Cells, 1:500-1:5000

Type: Primary
Antigen: MED20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty