Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP21 Rabbit Polyclonal Antibody

Supplier: Proteintech
55289-1-AP-150UL
10090-298EA 453 USD InStock
10090-298
Anti-MMP21 Rabbit Polyclonal Antibody
Antibodies
MMP21 belongs to the peptidase M10A family. It may have an important and specific function in tumor progression and embryogenesis. It cleaves alpha-1-antitrypsin. The proenzyme without the signal peptide has a calculated molecular mass of 62 kD; following activation by furin, the processed enzyme has a calculated molecular mass of 49 kD,and MMP21 protein was detected in fetal neuroectoderm, in placenta, and in leukocytes(OMIM).This antibody is specific to MMP21.

Western Blot: Mouse Liver Tissue, 1:200-1:2000; IHC: Human skin cancer Tissue, 1:20-1:200

Type: Primary
Antigen: MMP21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty