Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP26 Rabbit Polyclonal Antibody

Supplier: Proteintech
18087-1-AP-150UL
10090-302EA 453 USD InStock
10090-302
Anti-MMP26 Rabbit Polyclonal Antibody
Antibodies
MMP26(matrix metallopeptidase 26) may hydrolyze collagen type IV, fibronectin, fibrinogen, beta-casein, type I gelatin and alpha-1 proteinase inhibitor.MMP26 is expressed in placenta and uterus and is widely expressed in malignant tumors from different sources as well as in diverse tumor cell lines.The proenzyme without the signal peptide has a calculated molecular mass of 29 KDa and the processed enzyme has a calculated molecular mass of 19KDa.This antibody is specific to MMP29.

Western Blot:SKOV-3 Cells, 1:500-1:5000; IHC: Human prostate cancer, 1:20-1:200

Type: Primary
Antigen: MMP26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty