Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MUC16 Rabbit Polyclonal Antibody

Supplier: Proteintech
20077-1-AP-150UL
10090-596EA 453 USD InStock
10090-596
Anti-MUC16 Rabbit Polyclonal Antibody
Antibodies
MUC16, also named as CA125, is thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces. It is a serum marker that is used routinely in gynecologic practice to monitor patients with ovarian cancer. Most biochemical studies have concluded that MUC16 is a high molecular mass glycoprotein, although estimates of its size range from 200 to 2000 kDa with smaller “subunits” being described by some investigators. The antibody recognizes the C-term of MUC16.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: MUC16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty