Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NADK Rabbit Polyclonal Antibody

Supplier: Proteintech
15548-1-AP-150UL
10090-796EA 453 USD InStock
10090-796
Anti-NADK Rabbit Polyclonal Antibody
Antibodies
NAD kinase (NADK) is the sole NADP+-biosynthetic enzyme that catalyzes phosphorylation of NAD+ to yield NADP+ using ATP as a phosphoryl donor, and thus, plays a vital role in the cell and represents a potentially powerful antimicrobial drug target. It belongs to the NAD kinase family. The deduced 446-amino acid protein has a calculated molecular mass of 49.2 kD. The catalytically active human NADK is a homotetramer.

Western Blot: RAW 264.7 Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: NADK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty