Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NARF Rabbit Polyclonal Antibody

Supplier: Proteintech
12173-1-AP-150UL
10090-812EA 453 USD InStock
10090-812
Anti-NARF Rabbit Polyclonal Antibody
Antibodies
Prelamin A, is processed in the nucleus to lamin A by removal of its final 18 amino acids, including the cysteine residue in its C-terminal CAAX box, which is farnesylated. Nuclear prelamin A recognition factor (NARF) binds the farnesylated prelamin A C-terminal domain(preAct). NARF, at 52 kDa, also named as IOP2, is a small enough protein to freely diffuse through the nuclear pore complex and may be retained within the nucleus by binding nuclear proteins and it has all of the information necessary to be imported into the nucleus. It has 3 isoforms produced by alternative splicing with the molecular mass of 51 kDa, 56 kDa and 46 kDa,respectively.

Western Blot: Human Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: NARF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty