Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NEK9 Rabbit Polyclonal Antibody

Supplier: Proteintech
11192-1-AP-150UL
10091-020EA 453 USD InStock
10091-020
Anti-NEK9 Rabbit Polyclonal Antibody
Antibodies
NEK9 is also named as KIAA1995, NEK8, NERCC and belongs to the NEK Ser/Thr protein kinase family. It is originally identified as αβ-casein kinase that associates with a putative substrate Bicd2. It is also independently identified as a Nek6- and Ran GTPase-binding protein under a different name, Nercc1. Nek9, together with the highly similar (80% identical) Nek6 and Nek7, form a signaling module that is activated during mitosis and involved in the regulation of the mitotic spindle. The endogenous Nek9 (with predicted molecular mass of 120 kDa) has an apparent molecular mass of 600 kDa, also compatible with a tetramer.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: NEK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty