Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NIPSNAP3B Rabbit Polyclonal Antibody

Supplier: Proteintech
11789-1-AP-150UL
10091-116EA 453 USD InStock
10091-116
Anti-NIPSNAP3B Rabbit Polyclonal Antibody
Antibodies
NIPSNAP3B is a member of the NIPSNAP family with putative roles in vesicular trafficking . The gene of NIPSNAP3B maps to chromosome 9q31.3, and encodes a 247-amino-acid protein with a calculated molecular mass of 28 kDa. NIPSNAP3B gene has been reported to be highly associated with low bone mineral density (BMD) variation in postmenopausal Caucasian women .

Western Blot: Human Brain Tissue, 1:1000-1:10000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: NIPSNAP3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty