Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NKIAMRE Rabbit Polyclonal Antibody

Supplier: Proteintech
13888-1-AP-150UL
10091-128EA 453 USD InStock
10091-128
Anti-NKIAMRE Rabbit Polyclonal Antibody
Antibodies
NKIAMRE is also named as CDKL3(Cyclin-dependent kinase-like 3) and belongs to the protein kinase superfamily. It restricts cell growth or maintaining differentiation and may have a role in regulating cell growth and/or diferentiation. This protein contains the MAPK-like TDY motif for threonine and tyrosine phosphorylation, as well as the putative cyclin-binding motif. It has 2 isoforms produced by alternative splicing with the molecular mass of 52 kDa and 68 kDa.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: NKIAMRE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty