Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NMT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11546-1-AP-150UL
10091-168EA 453 USD InStock
10091-168
Anti-NMT1 Rabbit Polyclonal Antibody
Antibodies
NMT1 is a N-myristoyltransferase responsible for the transfer of myristate from CoA to an amino-terminal glycine of many eukaryotic proteins, which facilitates the targeting of proteins to membrane surfaces and is essential for viability of the organism. Insertional mutagenesis of the Nmt1 gene in Saccharomyces cerevisiae causes recessive lethality. Humans and mice possess two distinct but structurally similar enzymes, NMT1 and NMT2, ubiquitously expressed in most human and mouse tissues. Western analysis revealed that there are 4 isoforms of NMT1 with apparent molecular masses ranging from 49 to 68 kD. In cell fractionation studies, the 68-kD NMT1 isoform and NMT2 were present in both membrane and cytoplasmic fractions, while the smaller NMT1 isoforms were predominantly cytoplasmic.

Western Blot: Mouse pancreas Tissue, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: NMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty