Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PCYOX1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18868-1-AP-150UL
10091-960EA 453 USD InStock
10091-960
Anti-PCYOX1 Rabbit Polyclonal Antibody
Antibodies
PCYOX1(Prenylcysteine oxidase 1) is also named as KIAA0908, PCL1, prenylcysteine lyase and belongs to the prenylcysteine oxidase family. The deduced 505-amino acid protein contains a predicted signal sequence, several transmembrane regions, and four potential N-glycosylation sites. It is a glycosylated enzyme involved in prenylated protein catabolism, releasing a free cysteine and a hydrophobic isoprenoid product. Immunofluorescence studies of PCYOX1 showed a nonnuclear punctate pattern, and PCYOX1 colocalized with lysosomal marker LAMP1 in HEK293 cells. The enzyme is isolated from bovine brain membranes and exhibits an apparent molecular mass of 63 kDa.

IHC: Human Colon Tissue, 1:20-1:200

Type: Primary
Antigen: PCYOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty