Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDE8A Rabbit Polyclonal Antibody

Supplier: Proteintech
13956-1-AP-150UL
10092-016EA 453 USD InStock
10092-016
Anti-PDE8A Rabbit Polyclonal Antibody
Antibodies
PDE8A(high affinity cAMP-specific and IBMX-insensitive 3',5'-cyclic phosphodiesterase 8A) belongs to the cyclic nucleotide phosphodiesterase family and PDE8 subfamily. PDE8A plays a critical role in the modulation of at least one compartment of cAMP and hence PKA activity during β-adrenergic receptor (βAR) activation in ventricular myocytes. The full length protein contains several sites for protein phosphorylation, N-glycosylation, N-myristoylation, and amidation. It has 5 isoforms produced by alternative splicing with the molecular mass of 93 kDa, 88 kDa, 51 kDa and the isoform 4 and 5 have the same molecular mass of 66 kDa..

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200; IP: Mouse Testis Tissue, 1:500-1:5000

Type: Primary
Antigen: PDE8A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty