Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PIAS3 Rabbit Polyclonal Antibody

Supplier: Proteintech
13486-1-AP-150UL
10092-236EA 453 USD InStock
10092-236
Anti-PIAS3 Rabbit Polyclonal Antibody
Antibodies
PIAS3(E3 SUMO-protein ligase PIAS3) is a protein of 583 amino acid, has a molecular mass of 68 kDa and belongs to a family of proteins that includes sevsral other members. PIAS3 is an inhibitor of activated STAT3 and binds microphthalmia-associated transcription factor, which is a key DNA-binding protein, in rat basophilic leukemia cells and mouse melanocytes. The PIAS3 antibody detects PIAS3 (68 kDa) as well as its sumoylated form (85 kDa) in some normal mouse tissues, breast and brain tumor cell lines.

Western Blot: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: PIAS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty