Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PPP4R2 Rabbit Polyclonal Antibody

Supplier: Proteintech
22676-1-AP-150UL
10092-732EA 453 USD InStock
10092-732
Anti-PPP4R2 Rabbit Polyclonal Antibody
Antibodies
PPP4R2 is a regulatory subunit of protein phosphatase-4 (PPP4), a protein serine/threonine phosphatase implicated in microtubule organization at centrosomes and DNA double-strand break repair. The deduced 453-amino acid protein contains several potential phosphorylation sites and has a calculated molecular mass of 50.4 kD. Immunofluorescent analysis of HeLa and A431 cells revealed prominent localization of PPP4R2 at centrosomes, with weaker staining of cytoplasm and nucleus.

Western Blot:A549 Cells, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PPP4R2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty