Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TBX22 Rabbit Polyclonal Antibody

Supplier: Proteintech
11133-1-AP-150UL
10095-666EA 453 USD InStock
10095-666
Anti-TBX22 Rabbit Polyclonal Antibody
Antibodies
The frontonasal mass gives rise to the facial midline and fuses with the maxillary prominence to form the upper lip. T-box transcription factor 22 (Tbx22) belongs to the T-box family of transcription factors, which is essential for normal craniofacial development. Mutation in TBX22 in the frontnasal mass is reported in families with X-linked cleft palate and ankyloglossia .

  • Western Blot:PC-3 Cells, 1:200-1:2000

Type: Primary
Antigen: TBX22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty