Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UPB1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24603-1-AP-150UL
10096-802EA 453 USD InStock
10096-802
Anti-UPB1 Rabbit Polyclonal Antibody
Antibodies
BUP1(Beta-ureidopropionase) is also named as BUP1, Beta-alanine synthase, N-carbamoyl-beta-alanine amidohydrolase and belongs to the CN hydrolase family. It catalyzes the last step in the pyrimidine degradation pathway that it converts N-carbamyl-beta-aminoisobutyric acid and N-carbamyl-beta-alanine to, respectively, beta-aminoisobutyric acid and beta-alanine, ammonia and carbon dioxide. It is a protein with a calculated molecular mass of 43 kDa.

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: UPB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty